Hydrochloric acid 0.2 M analytical reagent
Catalog # DIUKHY14200-235
Supplier: DISTRIBUTOR
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Molarity:0.2 M
Specification Test Results
About this item
:
:36.46 g/mol
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324