Order Entry
United Kingdom
ContactUsLinkComponent
Methyl-4-bromobenzoate, Sigma-Aldrich®
Methyl-4-bromobenzoate, Sigma-Aldrich®
Catalog # SIAL407593-10G
Supplier:  Merck
CAS Number:  
Methyl-4-bromobenzoate, Sigma-Aldrich®
Catalog # SIAL407593-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :215.05 g/mol
Merck Index:13,01392
Boiling Pt:246 °C (1013 hPa)
Melting Pt:78…81 °C
Density:1.689 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:210-596-6
CAS Number:619-42-1
Formula:C₈H₇BrO₂
MDL Number:MFCD00013531