
1-Methylisatin 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2058-74-4
:
Formula:C₉H₇NO₂
MDL Number:MFCD00005812
:161.16 g/mol
Melting Pt:129…133 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-164-9
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Orange to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:129 to 133 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together