Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Piperidine

Glass bottle, 1 L
Catalog # MOLEM56236961
Flammable
Toxic
Corrosive
Marketsource
Supplier:   Molekula
Piperidine
Catalog # MOLEM56236961
Supplier:  Molekula
Marketsource
CAS Number:  110-89-4
Piperidine

Glass bottle, 1 L

CAS Number:  110-89-4
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexahydropyridine, Hexazane, Pentamethyleneimine
  • CAS Number:110-89-4
  • Formula:C₅H₁₁N
  • MDL Number:MFCD00005979
  • Molecular Weight:85.15 g/mol
  • Merck Index:12,07621
  • Boiling Pt:107.3 °C (1013 hPa)
  • Melting Pt:–10 °C
  • Density:0.8622 g/cm³ (20 °C)
  • Flash Pt:16 °C
  • Storage Temperature:Ambient
  • EINECS:203-813-0
  • UN:2401
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications