Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
N-Hydroxysuccinimide, Pierce™

N-Hydroxysuccinimide, Pierce™

Glass bottle for solids, 25 g
Catalog # PIER24500
N-Hydroxysuccinimide, Pierce™
Catalog # PIER24500
CAS Number:  6066-82-6
N-Hydroxysuccinimide, Pierce™
N-Hydroxysuccinimide, Pierce™

Glass bottle for solids, 25 g

CAS Number:  6066-82-6
Price
£82.60
Log in for availability
1 * 25 g
Product Details & Documents

Thermo Scientific Pierce NHS is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.

  • Related applications: Protein Labeling & Crosslinking
Synonyms:  1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
  • CAS Number:6066-82-6
  • Formula:C₄H₅NO₃
  • MDL Number:MFCD00005516
  • Molecular Weight:115.09 g/mol
  • Boiling Pt:363 °C (1013 hPa)
  • Melting Pt:93…95 °C
  • Storage Temperature:Refrigerator
  • EINECS:228-001-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.