N-Hydroxysuccinimide 98+%
: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
- :6066-82-6
- :C₄H₅NO₃
- :MFCD00005516
- :115.09 g/mol
- :363 °C (1013 hPa)
- :93…95 °C
- :Ambient
- :228-001-3
: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
- :6066-82-6
- :C₄H₅NO₃
- :MFCD00005516
- :115.09 g/mol
- :363 °C (1013 hPa)
- :93…95 °C
- :Ambient
- :228-001-3
- Appearance:White Crystalline powder or crystals (May have a tendency to cake)
- Infrared spectrum:Conforms
- Water:≤2.0 % (K.F.)
- Solubility:The product may contain some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone
- Titration with NaOH:98.0 to 101.0 % (On dried substance)
Product Family Options
- Pack typePk



