Order Entry
United Kingdom
ContactUsLinkComponent
2-Methyl-2-butene, Sigma-Aldrich®
2-Methyl-2-butene, Sigma-Aldrich®
  SIAL379263-800ML
 :  Merck
 :  
2-Methyl-2-butene, Sigma-Aldrich®
  SIAL379263-800ML
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    800 mL
  • Seq:
    2805
 :
 :70.13 g/mol
 :12,00649
 :39 °C (1013 hPa)
 :–124 °C
 :0.6623 g/cm³ (20 °C)
 :–45 °C
 :Refrigerator
 :208-156-3
 :2460
 :3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
 :GHS07
 :513-35-9
 :26760-64-5
 :C₅H₁₀
 :MFCD00009276