
AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-68-5
:
Formula:(CH₃)₂C(NH₂)CH₂OH
MDL Number:MFCD00008051
Merck Index:13,00448
Boiling Pt:165 °C (1013 hPa)
Melting Pt:18…26 °C
Density:0.667 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:67 °C
:89.14 g/mol
EINECS:204-709-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together