Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Isovaleric acid 99%

Isovaleric acid 99%

Corrosive
Isovaleric acid 99%
Certificates
Isovaleric acid 99%

3 Options Available Below

Product Details & Documents
Synonyms:  3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid
  • CAS Number:503-74-2
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00002726
  • Molecular Weight:102.13 g/mol
  • Merck Index:12,05251
  • Boiling Pt:176 °C (760 mmHg)
  • Melting Pt:–35 °C
  • Density:0.926 g/cm³
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • EINECS:207-975-3
  • UN:3265
  • ADR:8,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3990 to 1.4070 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific gravity:
    (25°C) 0.921 to 0.93
Product Family Options

Product Information