Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium methoxide
Sodium methoxide ≥98%
Sodium methoxide ≥98%
ABCRS93-1159.0500
:
abcr
:
Sodium methoxide ≥98%
ABCRS93-1159.0500
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
500 g
:
:
54.02 g/mol
:
127 °C
:
1.3 g/cm³ (20 °C)
:
204-699-5
:
1431
:
4.2,II
:
GHS02
:
GHS05
:
124-41-4
:
CH₃NaO
:
MFCD00012179