Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

Flammable
Irritant
   Sustainable Options Available
Marketsource
2-Propanol
   Sustainable Options Available
Marketsource
Certificates
2-Propanol

4 Options Available Below

Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BARRISOP005
  • BARRISOP025
  • BARRIPS1025
  • BARRISOP205
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    BARRISOP005
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    BARRISOP025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    BARRIPS1025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource
  • 205 L
    Log in for availability
    Log in for availability
    1 * 205 L
    1 * 205 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    BARRISOP205
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.