Acetic anhydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-24-7
:
Formula:(CH₃CO)₂O
MDL Number:MFCD00008705
Merck Index:13,00057
Boiling Pt:139.9 °C (1013 hPa)
Melting Pt:–73 °C
Density:1.08 g/cm³ (20 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:203-564-8
UN:1715
ADR:8,II
:M-Clarity™ quality level = MQ200
:102.09 g/mol
REACH:01-2119486470-36
:GHS02
:GHS05
:GHS07
Frequently Bought Together