Bis(2-butoxyethyl)ether 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-73-2
:
Formula:C₁₂H₂₆O₃
MDL Number:MFCD00009459
:218.34 g/mol
Boiling Pt:256 °C (760 mmHg)
Melting Pt:–60 °C
Density:0.88 g/cm³
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:204-001-9
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4225 to 1.4245 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤15 APHA
- Acidity (CH3COOH):≤0.01 %
- Peroxides:≤50 ppm (as H2O2)
Specification Test Results
Frequently Bought Together