Triethanolamine (trolamine) ≥99.0% analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:102-71-6
:
Formula:C₆H₁₅NO₃
MDL Number:MFCD00002855
:149.19 g/mol
Merck Index:13,09739
Boiling Pt:335.4 °C (1013 hPa)
Melting Pt:21.6 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:179 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-049-8
Specification Test Results
- Assay:Min. 99.0 %
- IR Spectrum:Passes test
- Density (20/4):1.120 - 1.130
- Solidification point:20 - 22 °C
- Diethanolamine:Max. 0.5 %
- Ethanolamine:Max. 0.1 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 50 ppm
- Water:Max. 0.2 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
Frequently Bought Together