Triethanolamine (trolamine), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:102-71-6
:
Formula:C₆H₁₅NO₃
MDL Number:MFCD00002855
:149.19 g/mol
Merck Index:13,09739
Boiling Pt:335.4 °C (1013 hPa)
Melting Pt:21.6 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:179 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-049-8
Frequently Bought Together