
Trifluoromethanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:421-83-0
:
Formula:CClF₃O₂S
MDL Number:MFCD00007451
:168.52 g/mol
Boiling Pt:32 °C (1013 hPa)
Density:1.583 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:207-009-0
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together