Perfluoro-tert-butanol 99%
Catalog # ACRO308390010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
About this item
Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol
:
:236.04 g/mol
EINECS:219-157-3
UN:2810
:GHS07
CAS Number:2378-02-1
Formula:C₄HF₉O
MDL Number:MFCD00042092
Boiling Pt:45 °C
Melting Pt:–17 °C
Density:1.69 g/cm³
ADR:6.1 II