1-Iodoperfluorbutane
Catalog # ABCRAB128074.0250
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
About this item
:
:345.93 g/mol
Boiling Pt:65 °C (1013 hPa)
Melting Pt:–2 °C
Density:2,0497 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-025-8
UN:3082
ADR:9,III
:GHS09
CAS Number:423-39-2
Formula:C₄F₉I
MDL Number:MFCD00001062