Order Entry
United Kingdom
ContactUsLinkComponent
Thiourea
Health
Irritant
Environment
Thiourea
Catalog # MOLE67805791-1KG
Supplier:  Molekula
CAS Number:  
undefined
Health
Irritant
Environment
Thiourea
Catalog # MOLE67805791-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :76.12 g/mol
Merck Index:13,09443
Melting Pt:174…177 °C
Density:1.435 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-543-5
UN:3077
ADR:9,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:62-56-6
Formula:NH₂CSNH₂
MDL Number:MFCD00008067