Citric acid
Glass bottle for solids, 25 g
Catalog # SIALC83155-25G
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
- CAS Number:77-92-9
- Formula:HOC(COOH)(CH₂COOH)₂
- MDL Number:MFCD00011669
- Molecular Weight:192.13 g/mol
- Merck Index:13,02350
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:149…151 °C
- Density:1.665 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-069-1
- REACH:01-2119457026-42-0000
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



