

Mercaptoacetic acid (thioglycolic acid) 98%
:68-11-1
:
:HSCH₂COOH
:MFCD00004876
:92.12 g/mol
:13,09410
:220 °C (760 mmHg)
:–16 °C
:1.325 g/cm³
:126 °C
:Refrigerator
:200-677-4
:1940
:8,II
:GHS05
:GHS06
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5030 to 1.5050 (20°C, 589 nm)
- Titration Iodimetric:≥97.5 %
- Iron (Fe):≤300 ppb
- Impurity:≤0.2 % Other acids (as TGA)
- UV:at 350 nm A: ≤0.01
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0003 meq/g
Frequently Bought Together