

Cobalt(II) acetate, anhydrous 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-48-7
:
Formula:C₄H₆CoO₄
MDL Number:MFCD00008689
:177.02 g/mol
Merck Index:14,2433
Melting Pt:298 °C (dec.)
Density:1.7755 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-755-8
UN:3077
ADR:9,III
:GHS08
:GHS07
Frequently Bought Together