


Mercury(II) acetate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1600-27-7
:
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165
:318.68 g/mol
Merck Index:13,05898
Melting Pt:179…182 °C
Density:3,27 g/cm³ (20 °C)
EINECS:216-491-1
UN:1629
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration Complexometric:≥99.0 %
- Residue after ignition:≤0.02 % (substances not reducible with formic acid)
- Chloride (Cl):≤50 ppm
- Nitrate (NO3):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
- Mercurous Mercury (as Hg):≤4000 ppm
Specification Test Results
Frequently Bought Together