Order Entry
United Kingdom
ContactUsLinkComponent
Ammonium formate, Sigma-Aldrich®
Irritant
Ammonium formate, Sigma-Aldrich®
Catalog # SIAL516961-100G
Supplier:  Merck
CAS Number:  
Irritant
Ammonium formate, Sigma-Aldrich®
Catalog # SIAL516961-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :63.06 g/mol
Merck Index:13,00525
Melting Pt:119…121 °C
Density:1.28 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
EINECS:208-753-9
 :GHS07
Storage Temperature:Ambient
CAS Number:540-69-2
Formula:HCO₂NH₄
MDL Number:MFCD00013103