Acetic acid glacial 100%, EMPROVE® EXPERT USP, BP, Ph. Eur., JP for biopharmaceutical production, SAFC®
: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
:64-19-7
:
:H₃CCOOH
:MFCD00036152
:118
:17
:1.05 g/cm³ (20 °C)
:60.05 g/mol
:38.5
:Ambient
:200-580-7
:2789
:8 (3)
:01-2119475328-30
:GHS02
:GHS05
- Environmentally Preferable:
- Assay (alkalimetric):99.8 - 100.5 %
- Identity:passes test
- Colour (Hazen):≤ 10 Hazen
- Appearance of the substance:passes test
- Formic acid, Formates and other oxidable impurities (as HCOOH):≤ 0.5 %
- Titratable base:≤ 0.0004 meq/g
- Solidification temperature:≥ 16.3 °C
- Chloride (Cl):≤ 2 ppm
- Sulfate (SO₄):≤ 5 ppm
- Heavy metals (as Pb):≤ 5 ppm
- Fe (Iron):≤ 0.2 ppm
- Other residual solvents (ICH Q3C):...excluded by production process
- Reducing substances (Ph Eur):passes test
- Substances reducing potassium permanganate:passes test
- Evaporation residue:≤ 50 ppm
- Dilution test:passes test
- Endotoxins:≤ 2.5 I.U./ml
Frequently Bought Together