Methyl formate, Supelco®
Catalog # 06547-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:60.05 g/mol
Merck Index:12,06153
Boiling Pt:32 °C (1013 hPa)
Melting Pt:–100.4 °C
Density:0.9713 g/cm³ (20 °C)
Flash Pt:–32 °C
EINECS:203-481-7
UN:1243
ADR:3,I
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:107-31-3
Formula:C₂H₄O₂
MDL Number:MFCD00003291