Methyl formate 97%, pure
:107-31-3
:
:C₂H₄O₂
:MFCD00003291
:60.05 g/mol
:12,06153
:31…33 °C (760 mmHg)
:–100 °C
:0.968 g/cm³
:–32 °C
:Ambient
:203-481-7
:1243
:3,I
:GHS07
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3420 to 1.3440 (20°C, 589 nm)
- Water:≤0.05 %
- GC:≥96.0 %
Frequently Bought Together