

Butyl formate
Catalog # EHERC13913300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:102.13 g/mol
Boiling Pt:107 °C (1013 hPa)
Melting Pt:–92 °C
Density:0.898 g/cm³ (20 °C)
Flash Pt:18 °C
Storage Temperature:Ambient
EINECS:209-772-5
UN:1128
ADR:3,II
:GHS02
:GHS07
CAS Number:592-84-7
Formula:C₅H₁₀O₂
MDL Number:MFCD00003296