

Propyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-60-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00009372
:102.13 g/mol
Merck Index:12,08026
Boiling Pt:102 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.886 g/cm³ (20 °C)
EINECS:203-686-1
UN:1276
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:10 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together