
Aluminium acetylacetonate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13963-57-0
:
Formula:C₁₅H₂₁AlO₆
MDL Number:MFCD00000013
Boiling Pt:315 °C (1013 hPa)
Melting Pt:192…195 °C
Density:1.204 g/cm³ (20 °C)
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:324.31 g/mol
EINECS:237-741-6
UN:3467
:GHS06
Specification Test Results
- Solubility:3 g/l (20 °C)
- Melting Point:190 - 193 °C
- Molar Mass:324.31 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:315 °C (1013 hPa)
- pH:8 (3 g/l, H2O, 20 °C)
- Ignition Point:500 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together