
Aluminium acetylacetonate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13963-57-0
:
Formula:C₁₅H₂₁AlO₆
MDL Number:MFCD00000013
Boiling Pt:315 °C (1013 hPa)
Melting Pt:190…195 °C
Density:1,204 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-741-6
UN:3467
ADR:6.1,II
:324.31 g/mol
:GHS06
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:190 to 195 °C
- Titration Complexometric:≥96.0 %
Specification Test Results
Frequently Bought Together