
Acetoin for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:513-86-0
:51555-24-9
Formula:C₄H₈O₂
MDL Number:MFCD00038696
:88.11 g/mol
Merck Index:13,00065
Boiling Pt:148 °C (1013 hPa)
Melting Pt:20 °C
Density:0.998 g/cm³ (20 °C)
UN:2621
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:208-174-1
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:20 °C Above 20°C conversion of dimer to monomer (15°C)
- Molar Mass:176.22 g/mol
- Boiling Point:147 - 148 °C
- Flash Point:45 °C
- Refractive Index:1.42 (20 °C, 589 nm)
- Density:1.012 g/cm3
Specification Test Results
Frequently Bought Together