Sodium pyruvate, Sigma-Aldrich®
Supplier: Merck
About this item
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Number:113-24-6
- Formula:CH₃COCOONa
- MDL Number:MFCD00002586
- Molecular Weight:110.04 g/mol
- Melting Pt:220…230 °C
- Density:1.718 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:204-024-4
3 Options Available Below
Product Details & Documents
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Number:113-24-6
- Formula:CH₃COCOONa
- MDL Number:MFCD00002586
- Molecular Weight:110.04 g/mol
- Melting Pt:220…230 °C
- Density:1.718 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:204-024-4
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SIALP8574-5G - Specifications:
More
Specifications
Cat. No.SIALP8574-25G - Specifications:
More
Specifications
Cat. No.SIALP8574-100G



