

Di-tert-butyl pyrocarbonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:24424-99-5
:
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805
:218.25 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:20…23 °C
Density:1.02 g/cm³ (20 °C)
Flash Pt:37 °C
UN:2930
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:246-240-1
:GHS02
:GHS06
Storage Temperature:Ambient
Frequently Bought Together