Ethyl pyruvate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
CAS Number:617-35-6
:
Formula:C₅H₈O₃
MDL Number:MFCD00009123
Boiling Pt:144…147 °C (1013 hPa)
Melting Pt:–50 °C
Density:1.05 g/cm³ (20 °C)
Molecular Weight:116.12 g/mol
Storage Temperature:Refrigerator
EINECS:210-511-2
ADR:3 III
:M-Clarity™ quality level = MQ200
UN:3272
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together