Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Acetone
Marketsource
Acetone
Marketsource
Acetone
Acetone
 :  2-Propanone, DMK, Propanone, Dimethyl ketone
  •  :67-64-1
  •  :CH₃COCH₃
  •  :MFCD00008765
  •  :58.08 g/mol
  •  :13,00067
  •  :56.2 °C (1013 hPa)
  •  :–95.4 °C
  •  :0.792 g/cm³ (20 °C)
  •  :<–20 °C (closed cup)
  •  :Ambient
  •  :200-662-2
  •  :1090
  •  :3,II
  •  :01-2119471330-49
  • Catalog No:
  • BARRACET200
  • BARRACET005
  • BARRACET010
  • BARRACET025
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
Product Family Options
  • Pack type
    Pk
  • 158 kg
    1 * 158 kg
    1 * 158 kg
      Safer Human and Environmental Health
    Marketsource
    Specifications
    Cat. No.
    BARRACET200
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
    Marketsource
  • Glass bottle
    5 L
    1 * 5 L
    1 * 5 L
      Safer Human and Environmental Health
    Marketsource
    Specifications
    Cat. No.
    BARRACET005
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
    Marketsource
  • 10 L
    1 * 10 L
    1 * 10 L
      Safer Human and Environmental Health
    Marketsource
    Specifications
    Cat. No.
    BARRACET010
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
    Marketsource
  • 25 L
    1 * 25 L
    1 * 25 L
      Safer Human and Environmental Health
    Marketsource
    Specifications
    Cat. No.
    BARRACET025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
    Marketsource