Order Entry
United Kingdom
ContactUsLinkComponent
 
Flammable
Irritant
1,2-Dichlorobutane, Sigma-Aldrich®
Catalog # SIAL105465-10ML
Supplier:  Merck
CAS Number:  
Flammable
Irritant
1,2-Dichlorobutane, Sigma-Aldrich®
Catalog # SIAL105465-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :127.01 g/mol
Boiling Pt:124.1 °C (1013 hPa)
Density:1.1116 g/cm³ (25 °C)
ADR:3,III
 :M-Clarity™ quality level = MQ100
EINECS:210-469-5
UN:1992
 :GHS02
 :GHS07
CAS Number:616-21-7
Formula:ClH₂CCHClCH₂CH₃
MDL Number:MFCD00013676