Sorbic acid, Sigma-Aldrich®
Catalog # SIAL82070-100G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:112.13 g/mol
Merck Index:13,08793
Boiling Pt:228 °C (1013 hPa)
Melting Pt:132…135 °C
Density:1.204 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-768-7
:GHS07
CAS Number:110-44-1
Formula:CH₃CH=CHCH=CHCOOH
MDL Number:MFCD00002703