Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
L-Norvaline

L-Norvaline

Glass bottle for solids, 5 g
Catalog # MOLEM34368263
Marketsource
Supplier:   Molekula
L-Norvaline
Catalog # MOLEM34368263
Supplier:  Molekula
Marketsource
CAS Number:  6600-40-4
L-Norvaline

Glass bottle for solids, 5 g

CAS Number:  6600-40-4
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
  • CAS Number:6600-40-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064421
  • Molecular Weight:117.15 g/mol
  • Merck Index:12,06813
  • Boiling Pt:228 °C (1013 hPa)
  • Melting Pt:101 °C
  • EINECS:229-543-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.