L-Norvaline 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
- CAS Number:6600-40-4
- Formula:C₅H₁₁NO₂
- MDL Number:MFCD00064421
- Molecular Weight:117.15 g/mol
- Merck Index:12,06813
- Boiling Pt:228 °C (1013 hPa)
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:229-543-3
3 Options Available Below
Product Details & Documents
Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
- CAS Number:6600-40-4
- Formula:C₅H₁₁NO₂
- MDL Number:MFCD00064421
- Molecular Weight:117.15 g/mol
- Merck Index:12,06813
- Boiling Pt:228 °C (1013 hPa)
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:229-543-3
Documents
ACRO22602
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless
- Specific optical rotation:+22.0° to +26.0° (20°C, 589 nm) (c=2, 5 N HCl)
- Titration with HClO4:98.5 to 101.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



