D-(-)-Mannitol for determination of boric acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:69-65-8
:
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287
:182.17 g/mol
Merck Index:13,05769
Boiling Pt:290…295 °C (4 hPa)
Melting Pt:164…169 °C
Density:1.49 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-711-8
Specification Test Results
- Solubility:213 g/l (25 °C)
- Melting Point:164 - 169 °C
- Molar Mass:182.17 g/mol
- Bulk Density:400 - 500 kg/m3
- Boiling Point:290 - 295 °C (4 hPa)
- Vapor Pressure:(20 °C)
- Density:1.49 g/cm3 (20 °C)
- pH:5 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:410 °C
- Viscosity kinematic:(20 °C) not applicable
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05983.1000_TDS_EN ProductPage_105983
Product specification
msds_105983
SDS
Frequently Bought Together