(R)-(-)-1,3-Butanediol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (R)-(-)-1,3-Dihydroxy butane, (R)-(-)-1,3-Butylene glykol, (R)-Butane-1,3-diol
CAS Number:6290-03-5
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00064277
Boiling Pt:203…204 °C
Density:1.005
Molecular Weight:90.12 g/mol
Flash Pt:121 °C (249 °F)
Storage Temperature:Ambient
EINECS:228-532-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together