Organic reference standard, Benzo[a]pyrene 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA20635000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:50-32-8
- Description:Benzo[a]pyrene 100 µg/ml in cyclohexane
- Formula:C₂₀H₁₂
- Description:Benzo[a]pyrene