Organic reference standard, Decachlorobiphenyl (PCB No. 209) 100 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, Decachlorobiphenyl (PCB No. 209) 100 µg/ml in isooctane, Dr Ehrenstorfer
- CAS No.:2051-24-3
- Formula:C₁₂Cl₁₀
- Description:Decachlorobiphenyl (PCB No. 209)
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Calcium 1.000 mg/l in water standard for ion chromatography
: VWR Chemicals
TETRASUL 1 * 50 mg
: Merck