L(-)-Malic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:97-67-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00064213
:134.09 g/mol
Merck Index:13,05730
Boiling Pt:322 °C (1013 hPa)
Melting Pt:101…104 °C
Density:1.595 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-601-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together