L(+)-Lactic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-33-4
:
Formula:C₃H₆O₃
MDL Number:MFCD00064266
:90.08 g/mol
Merck Index:13,05352
Boiling Pt:235 °C (1013 hPa)
Melting Pt:∼53 °C
Density:∼1.385 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-196-2
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together