Order Entry
United Kingdom
ContactUsLinkComponent
 
Organic reference standard, 2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20016000IO
 :  
Organic reference standard, 2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20016000IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    41411-62-5
  • Description:
    2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)