Order Entry
United Kingdom
ContactUsLinkComponent
Organic reference standard, 1-Bromo-4-fluorobenzene 10 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1-Bromo-4-fluorobenzene 10 µg/ml in methanol, Dr Ehrenstorfer
  EHERL10731000ME
 :  
Organic reference standard, 1-Bromo-4-fluorobenzene 10 µg/ml in methanol, Dr Ehrenstorfer
  EHERL10731000ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    460-00-4
  • Description:
    1-Bromo-4-fluorobenzene 10 µg/ml in methanol
  • Formula:
    C₆H₄BrF
  • Description:
    4-Bromofluorobenzene