

(S)-(-)-2-Bromopropionic acid 98%, >85% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32644-15-8
:
Formula:H₃CBrCHCOOH
MDL Number:MFCD00137325
:152.98 g/mol
Boiling Pt:203 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.7 g/cm³
Flash Pt:100 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4740 to 1.4760 (20°C, 589 nm)
- HPLC:≥97.5 %
- Enantiomeric excess:≥80 % (Typically)
- Specific optical rotation:-24° to -30° (20°C, 589 nm) (neat)
Specification Test Results
Frequently Bought Together