


Nickel(II) sulphate hexahydrate 98+% ACS
:10101-97-0
:
:NiSO₄·6H₂O
:MFCD00149813
:262.85 g/mol
:13,06541
:53 °C
:2,07 g/cm³ (20 °C)
:232-104-9
:3077
:9,III
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:Blue-green Crystals
- Titration Complexometric:98.0 to 102.0 %
- Chloride (Cl):≤0.001 %
- Calcium (Ca):≤0.005 %
- Cobalt (Co):≤0.002 %
- Copper (Cu):≤0.005 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.005 %
- Manganese (Mn):≤0.002 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.05 %
- Insoluble matter:≤0.005 %
- Nitrogen compounds (as N):≤0.002 %
Frequently Bought Together